accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OO(S)CH3Cl

CAS RN 73246-45-4

IUPAC Name: methyl (2S)-2-chloropropanoate
Synonyms: methyl (2S)-2-chloropropanoate
Molecular Weight (g/mol): 122.55
Molecular Formula: C4H7ClO2
InChi Key: JLEJCNOTNLZCHQ-VKHMYHEASA-N
SMILES: COC(=O)[C@H](C)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results