Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl (S)-(-)-2-Chloropropionate 98.0+%, TCI America™
SDP

Catalog No. C16345G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C16345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C16345G Supplier TCI America Supplier No. C16345G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 73246-45-4
Molecular Formula C4H7ClO2
Molecular Weight (g/mol) 122.55
MDL Number MFCD00062941
InChI Key JLEJCNOTNLZCHQ-VKHMYHEASA-N
Synonym (S)-(-)-2-Chloropropionic Acid Methyl Ester
PubChem CID 135535
IUPAC Name methyl (2S)-2-chloropropanoate
SMILES COC(=O)[C@H](C)Cl

Specifications

Color Yellow
Boiling Point 80°C
Quantity 5 g
UN Number 2933
Formula Weight 122.55
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl (S)-(-)-2-Chloropropionate
EINECSNumber (2)-3084
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.