accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOCH2

CAS RN 7328-17-8

IUPAC Name: 2-(2-ethoxyethoxy)ethyl prop-2-enoate
Synonyms: ethoxydiglycol acrylate
Molecular Weight (g/mol): 188.22
Molecular Formula: C9H16O4
InChi Key: FTALTLPZDVFJSS-UHFFFAOYSA-N
SMILES: CCOCCOCCOC(=O)C=C
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)