CAS RN 732938-37-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Syk Inhibitor IV, BAY 61-3606
The Syk Inhibitor IV, BAY 61-3606, also referenced under CAS 732938-37-8, controls the biological activity of Syk.