CAS RN 7331-52-4
IUPAC Name:
(5S)-5-hydroxyoxolan-2-one
Synonyms:
(5S)-5-hydroxyoxolan-2-one
Molecular Weight (g/mol):
102.09
Molecular Formula:
C4H6O3
InChi Key:
FYNVARJNCVQAAI-VKHMYHEASA-N
SMILES:
O[C@@H]1CCC(=O)O1
1
–
2
of
2
results
1
Sigma Aldrich (S)-(-)-3-Aminoquinuclidine Dihydrochloride
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.