Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-beta-Hydroxy-gamma-butyrolactone, 90+%, ee 99%, Thermo Scientific™

Catalog No. AAL2025606
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2025606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL2025606 Supplier Thermo Scientific Chemicals Supplier No. L2025606

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
(S)-(-)-β-Hydroxy-γ-butyrolactone acts as a chiral building block used in synthetic chemistry. It is also used in asymmetric synthesis.

Solubility
Miscible with water, alcohol and other organic solvent. Immiscible with light petroleum.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

CAS 7331-52-4
Density 1.302
Boiling Point 98°C to 100°C (0.3 mmHg)
Flash Point >110°C (230°F)
Molecular Formula C4H6O3
Refractive Index 1.4655
Quantity 5 g
MDL Number MFCD00211247
Beilstein 1280864
Synonym s-3-hydroxy-gamma-butyrolactone, s-4-hydroxydihydrofuran-2 3h-one, 4s-4-hydroxyoxolan-2-one, unii-2lie7314jh, s-4-hydroxytetrahydrofuran-2-one, s-beta-hydroxy-gamma-butyrolactone, 4s-4-hydroxydihydrofuran-2 3h-one, s-4,5-dihydro-4-hydroxy-2 3h-furanone, 2 3h-furanone, dihydro-4-hydroxy-, 4s, 4s-4-hydroxy-3,4,5-trihydrofuran-2-one
Solubility Information Miscible with water,alcohol and other organic solvent. Immiscible with light petroleum.
InChI Key FUDDLSHBRSNCBV-VKHMYHEASA-N
Optical Rotation −83° (c=2 in Ethanol)
SMILES C1C(COC1=O)O
IUPAC Name (4S)-4-hydroxyoxolan-2-one
Molecular Weight (g/mol) 102.089
PubChem CID 7269389
Formula Weight 102.09
Percent Purity ≥90%
Assay ee 99%
Chemical Name or Material (S)-(-)-beta-Hydroxy-gamma-butyrolactone
Show More Show Less
Hazard Category H317
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P272-P280g-P302+P352-P333+P313-P363-P501c
EINECSNumber 434-990-4
TSCA No
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.