accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7335-26-4

IUPAC Name: ethyl 2-methoxybenzoate
Synonyms: ethyl O-anisate
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: FNODWEPAWIJGPM-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=CC=C1OC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results