accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOONHNOO

CAS RN 734-25-8

IUPAC Name: 4-methyl-N-(4-nitrophenyl)benzene-1-sulfonamide
Synonyms: 4-methyl-N-(4-nitrophenyl)benzenesulfonamide
Molecular Weight (g/mol): 292.31
Molecular Formula: C13H12N2O4S
InChi Key: ACOIHAFYVPPSOZ-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)NC1=CC=C(C=C1)[N+]([O-])=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results