Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Nitro-p-toluenesulfonanilide, TCI America™
SDP

Catalog No. N063225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N063225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N063225G Supplier TCI America Supplier No. N063225G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 734-25-8
Molecular Formula C13H12N2O4S
Molecular Weight (g/mol) 292.309
MDL Number MFCD00129916
InChI Key ACOIHAFYVPPSOZ-UHFFFAOYSA-N
Synonym 4-Methyl-N-(4-nitrophenyl)benzenesulfonamide
PubChem CID 69772
IUPAC Name 4-methyl-N-(4-nitrophenyl)benzenesulfonamide
SMILES CC1=CC=C(C=C1)S(=O)(=O)NC2=CC=C(C=C2)[N+](=O)[O-]

Specifications

Melting Point 191°C
Quantity 25 g
Formula Weight 292.31
Physical Form Crystalline Powder
Chemical Name or Material 4′-Nitro-p-toluenesulfonanilide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.