accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7341-17-5

IUPAC Name: 2-ethylhexane-1-thiol
Synonyms: 2-ethylhexane-1-thiol
Molecular Weight (g/mol): 146.29
Molecular Formula: C8H18S
InChi Key: UCJMHYXRQZYNNL-UHFFFAOYNA-N
SMILES: CCCCC(CC)CS
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)