Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Ethylhexanethiol

Catalog No. 66914825G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
66-914-825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 66-914-825G Supplier Sigma Aldrich Supplier No. 66914825G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Ethylhexanethiol is an organothiol that forms a self-assembled monolayer (SAM) on a variety of surfaces such as metal oxides and metals. It modifies the surface by acting as a corrosion inhibitor and increases the stability of the surface.2-Ethylhexanethiol can be used as a capping agent on the gold surface for the development of chemiresistive vapor sensors. It also forms a SAM on copper indium sulfide (CuInS2) which can be used as an active layer on organic electronic devices such as organic light emitting diodes (OLEDs) and solar cells.

Specifications

CAS 7341-17-5
Density 0.843 g/mL (at 25°C)
Molecular Formula C8H18S
Refractive Index n20/D 1.457
Linear Formula C8H18S
MDL Number MFCD00039654
Quantity 25 g
Synonym 2-Ethyl-1-hexanethiol; 2-Ethylhexylmercaptan
Molecular Weight (g/mol) 146.29
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.