accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7342-41-8

IUPAC Name: 5'-(thiophen-2-yl)-[2,2'-bithiophene]-5-carbaldehyde
Synonyms: 5'-(thiophen-2-yl)-[2,2'-bithiophene]-5-carbaldehyde
Molecular Weight (g/mol): 276.39
Molecular Formula: C13H8OS3
InChi Key: PMPDDPJYARBNGV-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(S1)C1=CC=C(S1)C1=CC=CS1
Quantity 
  • (2)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results