Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-[5-(2-Thienyl)-2-thienyl]thiophene-2-carbaldehyde, Thermo Scientific™

Catalog No. SEW04053DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SEW04053DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SEW04053DA Supplier Thermo Scientific Chemicals Supplier No. SEW04053DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7342-41-8
Molecular Formula C13H8OS3
Molecular Weight (g/mol) 276.39
MDL Number MFCD00115181
InChI Key PMPDDPJYARBNGV-UHFFFAOYSA-N
Synonym 2,2':5',2-terthiophene-5-carboxaldehyde, 5-5-2-thienyl-2-thienyl thiophene-2-carbaldehyde, 2,2':5',2-terthiophene-5-carbaldehyde, 5-formyl-2,2':5',2-terthiophene, chembl91933, 5-5-thiophen-2-ylthiophen-2-yl thiophene-2-carbaldehyde, 5-5-thiophen-2-yl thiophen-2-yl thiophene-2-carbaldehyde, 2-formyl-alpha-terthienyl
PubChem CID 454742
SMILES O=CC1=CC=C(S1)C1=CC=C(S1)C1=CC=CS1

Specifications

Quantity 1 g
IUPAC Name 5'-(thiophen-2-yl)-[2,2'-bithiophene]-5-carbaldehyde
Formula Weight 276.4
Chemical Name or Material 5-[5-(2-Thienyl)-2-thienyl]thiophene-2-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.