accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHNH3CN

CAS RN 7343-34-2

IUPAC Name: 3,5-dimethyl-1H-1,2,4-triazole
Synonyms: 3,5-dimethyl-1H-1,2,4-triazole
Molecular Weight (g/mol): 97.12
Molecular Formula: C4H7N3
InChi Key: XYYXDARQOHWBPO-UHFFFAOYSA-N
SMILES: CC1=NC(C)=NN1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)