Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dimethyl-1,2,4-triazole 95.0+%, TCI America™
SDP

Catalog No. D44815G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D44815G 5 g
D44811G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D44815G Supplier TCI America Supplier No. D44815G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7343-34-2
Molecular Formula C4H7N3
Molecular Weight (g/mol) 97.121
MDL Number MFCD00656686
InChI Key XYYXDARQOHWBPO-UHFFFAOYSA-N
PubChem CID 139001
IUPAC Name 3,5-dimethyl-1H-1,2,4-triazole
SMILES CC1=NC(=NN1)C

Specifications

Melting Point 140°C
Color White-Yellow
Boiling Point 260°C
Quantity 5 g
Formula Weight 97.12
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dimethyl-1,2,4-triazole
EINECSNumber (5)-0600
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.