accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 73477-53-9

IUPAC Name: (2S)-2-[(diaminomethylidene)amino]pentanedioic acid
Synonyms: (2S)-2-[(diaminomethylidene)amino]pentanedioic acid
Molecular Weight (g/mol): 189.17
Molecular Formula: C6H11N3O4
InChi Key: RHVVRMJOHATSPD-VKHMYHEASA-N
SMILES: NC(N)=N[C@@H](CCC(O)=O)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results