Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-2-Guanidinoglutaric Acid 98.0+%, TCI America™
SDP

Catalog No. G0265100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G0265100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G0265100MG Supplier TCI America Supplier No. G0265100MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 73477-53-9
Molecular Formula C6H11N3O4
Molecular Weight (g/mol) 189.17
MDL Number MFCD00671536
InChI Key RHVVRMJOHATSPD-VKHMYHEASA-N
PubChem CID 135313
ChEBI CHEBI:7246
IUPAC Name (2S)-2-[(diaminomethylidene)amino]pentanedioic acid
SMILES NC(N)=N[C@@H](CCC(O)=O)C(O)=O

Specifications

Color White
Quantity 100 mg
Formula Weight 189.17
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(-)-2-Guanidinoglutaric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.