accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOH3COOH3COOH3COORhRh

CAS RN 73482-96-9

IUPAC Name: bis(λ²-rhodium(2+)) tetraoctanoate
Synonyms: bis(λ²-rhodium(2+)) tetraoctanoate
Molecular Weight (g/mol): 778.64
Molecular Formula: C32H60O8Rh2
InChi Key: FZXFNYFVNKTQSX-UHFFFAOYSA-J
SMILES: [Rh++].[Rh++].CCCCCCCC([O-])=O.CCCCCCCC([O-])=O.CCCCCCCC([O-])=O.CCCCCCCC([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results