Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rhodium(II) Octanoate Dimer, TCI America™
SDP

Catalog No. R0161200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R0161200MG 200 mg
R01611G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R0161200MG Supplier TCI America Supplier No. R0161200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 73482-96-9
Molecular Formula C32H60O8Rh2
Molecular Weight (g/mol) 778.635
MDL Number MFCD00064724
InChI Key FZXFNYFVNKTQSX-UHFFFAOYSA-J
Synonym chloro 1,2,3-eta-3-phenyl-2-propenyl 1,3-bis 2,6-di-i-propylphenyl imidazol-2-ylidene palladium ii
PubChem CID 131858392
IUPAC Name octanoate;rhodium(2+)
SMILES CCCCCCCC(=O)[O-].CCCCCCCC(=O)[O-].CCCCCCCC(=O)[O-].CCCCCCCC(=O)[O-].[Rh+2].[Rh+2]

Specifications

Color Green
Quantity 200 mg
Formula Weight 778.64
Physical Form Crystalline Powder
Chemical Name or Material Rhodium(II) Octanoate Dimer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.