accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OClN

CAS RN 73568-26-0

IUPAC Name: 2-chloro-8-methylquinoline-3-carbaldehyde
Synonyms: 2-chloro-8-methylquinoline-3-carbaldehyde
Molecular Weight (g/mol): 205.64
Molecular Formula: C11H8ClNO
InChi Key: YPBRSXNRWFUUOE-UHFFFAOYSA-N
SMILES: CC1=C2N=C(Cl)C(C=O)=CC2=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results