Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-8-methylquinoline-3-carboxaldehyde 98.0+%, TCI America™
SDP

Catalog No. C31095G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C31095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C31095G Supplier TCI America Supplier No. C31095G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

Specifications

CAS 73568-26-0
Melting Point 139°C
Molecular Formula C11H8ClNO
MDL Number MFCD02227041
Quantity 5 g
Synonym 2-chloro-8-methyl-3-quinoline carboxaldehyde, 2-chloro-8-methylquinoline-3-carboxaldehyde, 2-chloro-8-methyl-3-quinolinecarbaldehyde, 2-chloro-8-methyl-quinoline-3-carbaldehyde, acmc-1cokl, 2-chloro-3-formyl-8-methylquinoline, 2-chloro-8-methylquinoline 3-carbaldehyde, 2-chloro-8-methyl quinoline-3-carbaldehyde, 2-chloro-8-methyl-3-quinoline carbaldehyde, 2-chloro-8-methylquinoline 3-carbalde-hyde
InChI Key YPBRSXNRWFUUOE-UHFFFAOYSA-N
SMILES CC1=CC=CC2=CC(=C(N=C12)Cl)C=O
IUPAC Name 2-chloro-8-methylquinoline-3-carbaldehyde
Molecular Weight (g/mol) 205.641
PubChem CID 689082
Formula Weight 205.64
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-8-methylquinoline-3-carboxaldehyde
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.