CAS RN 7361-31-1
IUPAC Name:
dihydrogen methyl (2R)-2-amino-3-(4-hydroxyphenyl)-2-methylpropanoate methyl (2S)-2-amino-3-(4-hydroxyphenyl)-2-methylpropanoate dichloride
Synonyms:
dihydrogen methyl (2R)-2-amino-3-(4-hydroxyphenyl)-2-methylpropanoate methyl (2S)-2-amino-3-(4-hydroxyphenyl)-2-methylpropanoate dichloride
Molecular Weight (g/mol):
491.41
Molecular Formula:
C22H32Cl2N2O6
InChi Key:
LBSDGCNTBFGZKY-VPLDNDRNSA-N
SMILES:
[H+].[H+].[Cl-].[Cl-].COC(=O)[C@](C)(N)CC1=CC=C(O)C=C1.COC(=O)[C@@](C)(N)CC1=CC=C(O)C=C1
1
–
1
of
1
results
1
MP Biomedicals, Inc DL-a-Methyl-P-Tyrosine Methylester Hydrocloride, MP Biomedicals™
CAS: 7361-31-1 Molecular Formula: C22H32Cl2N2O6 Molecular Weight (g/mol): 491.41 MDL Number: MFCD00012606 InChI Key: LBSDGCNTBFGZKY-VPLDNDRNSA-N Synonym: α-Methyl-DL-tyrosine methyl ester hydrochloride,α-MT methyl ester hydrochloride,2-Methyl-4-hydroxy-DL-phenylalanine methyl ester hydrochloride,AMPT