Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-a-Methyl-P-Tyrosine Methylester Hydrocloride, MP Biomedicals™

Catalog No. MP522780801 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP522780801 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP522780801 Supplier MP Biomedicals, Inc Supplier No. 0522780801

DL-a-Methyl-P-Tyrosine Methylester Hydrocloride

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7361-31-1
Molecular Formula C22H32Cl2N2O6
Molecular Weight (g/mol) 491.41
MDL Number MFCD00012606
InChI Key LBSDGCNTBFGZKY-VPLDNDRNSA-N
Synonym α-Methyl-DL-tyrosine methyl ester hydrochloride, α-MT methyl ester hydrochloride, 2-Methyl-4-hydroxy-DL-phenylalanine methyl ester hydrochloride, AMPT

Specifications

Melting Point 192°C (decomposition)(literature)
Quantity 1 g
Solubility Information DMSO: ≥20mg/mL, clear, colorless to faintly yellow
RTECSNumber YP2900000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.