accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7367-82-0

H3COOCH3(E)

CAS RN 7367-82-0

IUPAC Name: ethyl (2E)-oct-2-enoate
Synonyms: ethyl 2-octenoate
Molecular Weight (g/mol): 170.25
Molecular Formula: C10H18O2
InChi Key: AISZSTYLOVXFII-CMDGGOBGSA-N
SMILES: CCCCC\C=C\C(=O)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results