Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl trans-2-Octenoate 96.0+%, TCI America™
SDP

Catalog No. O028625ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O028625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O028625ML Supplier TCI America Supplier No. O028625ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7367-82-0
Molecular Formula C10H18O2
Molecular Weight (g/mol) 170.252
MDL Number MFCD00015287
InChI Key AISZSTYLOVXFII-CMDGGOBGSA-N
Synonym ethyl trans-2-octenoate, ethyl 2-octenoate, ethyl e-2-octenoate, ethyl e-oct-2-enoate, 2-octenoic acid, ethyl ester, ethyl 2-octenoate, trans, unii-44bpd919tx, 2-octenoic acid, ethyl ester, e, fema no. 3643, ethyl oct-2-enoate
PubChem CID 5364399
IUPAC Name ethyl (E)-oct-2-enoate
SMILES CCCCCC=CC(=O)OCC

Specifications

Color Colorless
Boiling Point 69°C
Quantity 25 mL
Formula Weight 170.25
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl trans-2-Octenoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.