accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HNOH2NOHO

CAS RN 7369-76-8

IUPAC Name: 2-(2-aminoacetamido)butanoic acid
Synonyms: 2-(2-aminoacetamido)butanoic acid
Molecular Weight (g/mol): 160.17
Molecular Formula: C6H12N2O3
InChi Key: XHUWGEILXFVFMR-UHFFFAOYNA-N
SMILES: CCC(NC(=O)CN)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results