accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH

CAS RN 737-89-3

IUPAC Name: N-(naphthalen-1-yl)naphthalen-1-amine
Synonyms: di-naphthalen-1-yl-amine
Molecular Weight (g/mol): 269.35
Molecular Formula: C20H15N
InChi Key: VMVGVGMRBKYIGN-UHFFFAOYSA-N
SMILES: N(C1=C2C=CC=CC2=CC=C1)C1=C2C=CC=CC2=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results