accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 737000-84-9

IUPAC Name: 1-isothiocyanato-2,3-dihydro-1H-indene
Synonyms: 1-isothiocyanato-2,3-dihydro-1H-indene
Molecular Weight (g/mol): 175.25
Molecular Formula: C10H9NS
InChi Key: YQGGEOQEFFXRLO-UHFFFAOYNA-N
SMILES: S=C=NC1CCC2=CC=CC=C12
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)