Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-1-Indanyl isothiocyanate, 95%, Thermo Scientific™

Catalog No. AAL1997903
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1997903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1997903 Supplier Thermo Scientific Chemicals Supplier No. L1997903

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 737000-84-9
Molecular Formula C10H9NS
Molecular Weight (g/mol) 175.249
MDL Number MFCD04039331
InChI Key YQGGEOQEFFXRLO-JTQLQIEISA-N
Synonym s-+-1-indanyl isothiocyanate, 1s-1-isothiocyanato-2,3-dihydro-1h-indene, s-1-isothiocyanato-2,3-dihydro-1h-indene
PubChem CID 7016322
IUPAC Name (1S)-1-isothiocyanato-2,3-dihydro-1H-indene
SMILES C1CC2=CC=CC=C2C1N=C=S

Specifications

Boiling Point 108°C to 113°C (0.1 mmHg)
Quantity 1 g
UN Number UN2922
Formula Weight 175.25
Percent Purity 95%
Chemical Name or Material (S)-(+)-1-Indanyl isothiocyanate
Hazard Category Danger
Hazard Statement GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
Recommended Storage 2° to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.