accessibility menu, dialog, popup

UserName

Viewing profile as user:
SSOOOHONH(R)CH3H3CCH3

CAS RN 73724-43-3

IUPAC Name: (2R)-3-(tert-butyldisulfanyl)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Synonyms: (2R)-3-(tert-butyldisulfanyl)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid
Molecular Weight (g/mol): 431.57
Molecular Formula: C22H25NO4S2
InChi Key: ZDUMTHLUTJOUML-IBGZPJMESA-N
SMILES: CC(C)(C)SSC[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)