accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOOHONH(S)

CAS RN 73724-46-6

IUPAC Name: benzyl (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-hydroxypropanoate
Synonyms: benzyl (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-hydroxypropanoate
Molecular Weight (g/mol): 417.46
Molecular Formula: C25H23NO5
InChi Key: GXJVEJVEJKIXCH-QHCPKHFHSA-N
SMILES: OC[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(=O)OCC1=CC=CC=C1
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)