accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHNI

CAS RN 73746-45-9

IUPAC Name: 5-iodo-2-methyl-1H-imidazole
Synonyms: 4-iodo-2-methyl-3H-imidazole
Molecular Weight (g/mol): 208
Molecular Formula: C4H5IN2
InChi Key: SEDSLMWYUQACGR-UHFFFAOYSA-N
SMILES: CC1=NC=C(I)N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results