Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-iodo-2-methyl-1H-imidazole, Thermo Scientific™

Catalog No. MO00831EA
Change view
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MO00831EA 10 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MO00831EA Supplier Thermo Scientific Chemicals Supplier No. MO00831EA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 73746-45-9
Molecular Formula C4H5IN2
Molecular Weight (g/mol) 208.00
MDL Number MFCD02684335
InChI Key SEDSLMWYUQACGR-UHFFFAOYSA-N
PubChem CID 2794623
IUPAC Name 5-iodo-2-methyl-1H-imidazole
SMILES CC1=NC=C(I)N1

Specifications

Packaging Amber Glass Bottle
Quantity 10 g
Synonym 4-iodo-2-methyl-1h-imidazole, 2-methyl-4 5-iodo-1 h-imidazole, 4-iodo-2-methylimidazole, 2-methyl-4-iodo-1h-imidazole, 4-iodo-2-methyl-3h-imidazole, 1h-imidazole, 4-iodo-2-methyl, 2-methyl-4-jodimidazol, ksc638k7d, 1h-imidazole,4-iodo-2-methyl, 1h-imidazole,5-iodo-2-methyl
Formula Weight 208.0016
Chemical Name or Material 4-iodo-2-methyl-1H-imidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.