accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7376-03-6

IUPAC Name: phenyl(pyren-1-yl)methanone
Synonyms: phenyl(pyren-1-yl)methanone
Molecular Weight (g/mol): 306.36
Molecular Formula: C23H14O
InChi Key: FGCOCPAQFSREBF-UHFFFAOYSA-N
SMILES: O=C(C1=CC=CC=C1)C1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)