Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzoylpyrene, 98%, Thermo Scientific™

Catalog No. AC449310010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC449310010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC449310010 Supplier Thermo Scientific Chemicals Supplier No. 449310010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7376-03-6
Molecular Formula C10H11N2O3
Molecular Weight (g/mol) 207.21
InChI Key VHAXWROFYVPXMZ-UHFFFAOYSA-M
Synonym n-4-aminobenzoyl-beta-alanine, 3-4-amino-benzoylamino-propionic acid, 3-4-aminobenzamido propanoic acid, 4-aminobenzoyl-beta-alanine, unii-ab85ljq945, 3-4-aminobenzoyl amino propanoic acid, n-4-aminophenyl carbonyl-beta-alanine, 3-4-aminophenyl formamido propanoic acid, 3-4-aminophenyl carbonylamino propanoic acid
PubChem CID 719629
IUPAC Name 3-[(4-aminobenzoyl)amino]propanoic acid
SMILES NC1=CC=C(C=C1)C(=O)NCCC([O-])=O

Specifications

Melting Point 127.0°C to 129.0°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Solubility Information Solubility in water: insoluble
Quantity 1 g
Formula Weight 306.36
Percent Purity 98%
Chemical Name or Material 1-Benzoylpyrene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.