accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCH3CH3

CAS RN 7379-12-6

IUPAC Name: 2-methylhexan-3-one
Synonyms: 2-methyl-3-hexanone
Molecular Weight (g/mol): 114.19
Molecular Formula: C7H14O
InChi Key: HIGGFWFRAWSMBR-UHFFFAOYSA-N
SMILES: CCCC(=O)C(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results