Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-3-hexanone, TCI America™
SDP

Catalog No. M057010G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M057010G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M057010G Supplier TCI America Supplier No. M057010G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7379-12-6
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.19
MDL Number MFCD00009399
InChI Key HIGGFWFRAWSMBR-UHFFFAOYSA-N
Synonym 2-methyl-3-hexanone, 3-hexanone, 2-methyl, isopropyl propyl ketone, 2-methyl-hexan-3-one, propyl isopropyl ketone, isopropyl-n-propyl ketone, 4-01-00-03328 beilstein handbook reference
PubChem CID 23847
IUPAC Name 2-methylhexan-3-one
SMILES CCCC(=O)C(C)C

Specifications

Color Yellow
Boiling Point 136°C
UN Number 1224
Quantity 10 g
Formula Weight 114.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyl-3-hexanone
RTECSNumber MP3900000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.