accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 73821-97-3

CH3CH3CH3OOHNOOOHO

CAS RN 73821-97-3

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-5-(cyclohexyloxy)-5-oxopentanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]-5-(cyclohexyloxy)-5-oxopentanoic acid
Molecular Weight (g/mol): 329.39
Molecular Formula: C16H27NO6
InChi Key: FDNMLANBNJDIRG-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CCC(=O)OC1CCCCC1)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)