Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Cyclohexyl N-(tert-Butoxycarbonyl)-L-glutamate 94.0+%, TCI America™
SDP

Catalog No. B21795G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21795G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B21795G Supplier TCI America Supplier No. B21795G

Chemical Identifiers

CAS 73821-97-3
Molecular Formula C16H27NO6
Molecular Weight (g/mol) 329.393
MDL Number MFCD00065570
InChI Key FDNMLANBNJDIRG-LBPRGKRZSA-N
Synonym boc-glu ochex-oh, boc-glu ochx-oh, s-2-tert-butoxycarbonyl amino-5-cyclohexyloxy-5-oxopentanoic acid, boc-l-glutamic acid 5-cyclohexyl ester, n-boc-l-glutamic acid 5-cyclohexyl ester, n-tert-butoxycarbonyl-l-glutamic acid 5-cyclohexyl ester, 2s-2-tert-butoxycarbonyl amino-5-cyclohexyloxy-5-oxopentanoic acid, boc-glu ochx, pubchem12179, boc-l-glutamicacid-5-cyclohexylester
PubChem CID 7017890
IUPAC Name (2S)-5-cyclohexyloxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxopentanoic acid
SMILES CC(C)(C)OC(=O)NC(CCC(=O)OC1CCCCC1)C(=O)O

Specifications

Melting Point 57°C
Color White
Quantity 5 g
Formula Weight 329.39
Percent Purity ≥94.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Cyclohexyl N-(tert-Butoxycarbonyl)-L-glutamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.