accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7386-21-2

IUPAC Name: 2-(4-chlorophenyl)-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: 2-(4-chlorophenyl)isoindole-1,3-dione
Molecular Weight (g/mol): 257.67
Molecular Formula: C14H8ClNO2
InChi Key: QKHKQJWODBAIMN-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)N1C(=O)C2=CC=CC=C2C1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results