accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 73896-36-3

IUPAC Name: 6-methoxy-3-nitropyridin-2-amine
Synonyms: 6-methoxy-3-nitropyridin-2-amine
Molecular Weight (g/mol): 169.14
Molecular Formula: C6H7N3O3
InChi Key: RDJILYVRVOTMTQ-UHFFFAOYSA-N
SMILES: COC1=NC(N)=C(C=C1)[N+]([O-])=O
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results