Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-6-methoxy-3-nitropyridine 97.0+%, TCI America™
SDP

Catalog No. A23725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23725G 5 g
A237225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A23725G Supplier TCI America Supplier No. A23725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 73896-36-3
Molecular Formula C6H7N3O3
Molecular Weight (g/mol) 169.14
MDL Number MFCD03206481
InChI Key RDJILYVRVOTMTQ-UHFFFAOYSA-N
Synonym 2-amino-6-methoxy-3-nitropyridine, 2-amino-3-nitro-6-methoxypyridine, 6-methoxy-3-nitro-2-pyridinamine, 2-amino-3-nitro-6-methoxy pyridine, 2-pyridinamine, 6-methoxy-3-nitro, 6-methoxy-3-nitro-2-pyridylamine, 6-methoxy-3-nitro-pyridin-2-ylamine, pubchem5689, acmc-1bcyf, ksc496m1p
PubChem CID 3852428
IUPAC Name 6-methoxy-3-nitropyridin-2-amine
SMILES COC1=NC(=C(C=C1)[N+](=O)[O-])N

Specifications

Melting Point 171°C
Quantity 5 g
Formula Weight 169.14
Percent Purity ≥97.0% (GC)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-6-methoxy-3-nitropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.