CAS RN 739364-95-5
IUPAC Name:
2-methylpyrazolo[1,5-a]pyrimidine-6-carboxylic acid
Synonyms:
2-methylpyrazolo[1,5-a]pyrimidine-6-carboxylic acid
Molecular Weight (g/mol):
177.16
Molecular Formula:
C8H7N3O2
InChi Key:
YHVGUXFUOSJCFJ-UHFFFAOYSA-N
SMILES:
CC1=NN2C=C(C=NC2=C1)C(O)=O