accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 73956-15-7

IUPAC Name: 2,2-diethoxyethanethioamide
Synonyms: 2,2-diethoxyethanethioamide
Molecular Weight (g/mol): 163.24
Molecular Formula: C6H13NO2S
InChi Key: MQSDGAKLSVITHP-UHFFFAOYSA-N
SMILES: CCOC(OCC)C(N)=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results