Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Diethoxyethanethioamide, 97%, Thermo Scientific™

Catalog No. MO01292DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MO01292DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MO01292DA Supplier Thermo Scientific Chemicals Supplier No. MO01292DA

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 73956-15-7
Molecular Formula C6H13NO2S
Molecular Weight (g/mol) 163.235
MDL Number MFCD06658986
InChI Key MQSDGAKLSVITHP-UHFFFAOYSA-N
PubChem CID 2794734
IUPAC Name 2,2-diethoxyethanethioamide
SMILES CCOC(C(=S)N)OCC

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 2,2-diethoxy-ethanethioamide, diethoxythioacetamide, 2,2-diethoxythioacetamide, 2,2,-diethoxyethanethioamide, ethanethioamide, 2,2-diethoxy, ethanethioamide,2,2-diethoxy, 2,2-bis ethyloxy ethanethioamide, 1-amino-2,2-diethoxyethane-1-thione
Formula Weight 163.24
Percent Purity 97%
Chemical Name or Material 2, 2-Diethoxyethanethioamide
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.