accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClCH3NCH3CH2H2C

CAS RN 7398-69-8

IUPAC Name: dimethylbis(prop-2-en-1-yl)azanium chloride
Synonyms: PolyDADMAC polyquaternium-6
Molecular Weight (g/mol): 161.67
Molecular Formula: C8H16ClN
InChi Key: GQOKIYDTHHZSCJ-UHFFFAOYSA-M
SMILES: [Cl-].C[N+](C)(CC=C)CC=C
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results