Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diallyldimethylammonium chloride solution

Catalog No. 3482793L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 L
3 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-827-93L 3 L
34-827-91L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 34-827-93L Supplier Sigma Aldrich Supplier No. 3482793L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Diallyldimethylammonium chloride solution (DADMAC) is a hydrophilic quaternary ammonium compound that can be dissolved in an aqueous solution as a positively charged colloid.DADMAC is used as a cationic monomer solution for the fabrication of ion-selective polyelectrolytic anodized aluminium oxide (AAO) membranes which can be used for the development of electrical power generation systems. It may be grafted on carboxymethyl cellulose (CMC) for use as an absorbent for cationic dyes.

Specifications

CAS 7398-69-8
Molecular Formula C8H16ClN
Linear Formula [(CH3)2 N+(CH2CH=CH2)2 ]Cl-
Quantity 3 L
MDL Number MFCD00043200
Synonym Dimethyldiallylammonium chloride
Molecular Weight (g/mol) 161.67
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.