accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3N

CAS RN 7399-50-0

IUPAC Name: 2-(pentan-3-yl)pyridine
Synonyms: 2-(1-ethylpropyl)pyridine
Molecular Weight (g/mol): 149.24
Molecular Formula: C10H15N
InChi Key: FTYAXYWEDPWJCJ-UHFFFAOYSA-N
SMILES: CCC(CC)C1=CC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results