accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NH2NNH2OHO

CAS RN 74-79-3

IUPAC Name: 2-amino-5-[(diaminomethylidene)amino]pentanoic acid
Synonyms: 2-amino-5-[(diaminomethylidene)amino]pentanoic acid
Molecular Weight (g/mol): 174.2
Molecular Formula: C6H14N4O2
InChi Key: ODKSFYDXXFIFQN-UHFFFAOYNA-N
SMILES: NC(CCCN=C(N)N)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (33)
  • (1)
Quantity 
  • (7)
  • (1)
  • (9)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (4)
  • (4)
Percent Purity 
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (1)
  • (14)
  • (1)
Grade 
  • (5)
  • (1)
  • (3)
  • (1)
  • (1)
  • (5)
  • (7)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (23)
  • (17)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (6)
  • (16)
  • (7)
  • (2)
  • (13)
  • (1)
  • (2)
  • (33)
  • (1)
  • (7)
  • (1)
  • (9)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (4)
  • (4)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (1)
  • (14)
  • (1)
  • (5)
  • (1)
  • (3)
  • (1)
  • (1)
  • (5)
  • (7)
  • (2)