Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medisca Arginine (L), USP
SDP

Catalog No. 14185638 Shop All Medisca Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-185-638 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-185-638 Supplier Medisca Supplier No. 050208
Get 1 Extra Chemical When You Buy 4
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Arginine (L), USP comes as white, practically odorless crystals. It is freely soluble in water and sparingly soluble in alcohol.

Arginine (L), USP comes as white, practically odorless crystals. It is freely soluble in water and sparingly soluble in alcohol.

Alternative name(s): Arginine, L-Arginine, L-Arg

Specifications

Chemical Name or Material Arginine (L), USP
Packaging Jar, 1000mL, White, HDPE, Round, 89-400, TE Pano Neck
CAS 74-79-3
Type Drug Standard
Molecular Weight (g/mol) 174.203
Quantity 500 g
Percent Purity 98.5 to 101.5 %
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.